APO455832310 Apollo Scientific Ltd

3-Bromo-N-cyclopropyl-2-nitroaniline

  • MDL number MFCD22415284
  • Cas Number : 1396554-46-3
  • Molecular Weight : 257.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-N-cyclopropyl-2-nitroaniline
Inchi Code 1S/C9H9BrN2O2/c10-7-2-1-3-8(9(7)12(13)14)11-6-4-5-6/h1-3,6,11H,4-5H2
Inchi Key IHVDMCLNJSTNHO-UHFFFAOYSA-N

Description

Synonym 2-Bromo-6-(cyclopropylamino)nitrobenzene, N-(3-Bromo-2-nitrophenyl)cyclopropylamine
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-n-cyclopropyl-2-nitroaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-n-cyclopropyl-2-nitroaniline
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart