APO455832313 Apollo Scientific Ltd

3-Acetyl-2-chloropyridine

  • MDL number MFCD03840751
  • Cas Number : 55676-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Chloropyridin-3-yl)ethan-1-one, 2-Chloro-3-ethanoylpyridine
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-acetyl-2-chloropyridine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-acetyl-2-chloropyridine
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart