APO455832315 Apollo Scientific Ltd

2-Bromo-N4-methylpyridine-3,4-diamine

  • MDL number MFCD22415287
  • Cas Number : 1396554-44-1
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-N~4~-methyl-3,4-pyridinediamine
Inchi Code 1S/C6H8BrN3/c1-9-4-2-3-10-6(7)5(4)8/h2-3H,8H2,1H3,(H,9,10)
Inchi Key IRUUOKUGVROVDG-UHFFFAOYSA-N

Description

Synonym 3-Amino-2-bromo-4-(methylamino)pyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-n4-methylpyridine-34-diamine

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/2-bromo-n4-methylpyridine-34-diamine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart