APO455832317 Apollo Scientific Ltd

2,3-Dichloro-4-hydroxybenzaldehyde

  • MDL number MFCD07775189
  • Cas Number : 16861-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Dichloro-4-formylphenol
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31663

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31663
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart