APO455832330 Apollo Scientific Ltd

Quinoline-6-carboxaldehyde

  • MDL number MFCD00805836
  • Cas Number : 4113-04-6
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 75-76
Iupac Name 6-quinolinecarbaldehyde
Inchi Code 1S/C10H7NO/c12-7-8-3-4-10-9(6-8)2-1-5-11-10/h1-7H
Inchi Key VUAOIXANWIFYCU-UHFFFAOYSA-N

Description

Synonym 6-Formylquinoline, 6-Formyl-1-azanaphthalene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 75-76
Product Link https://store.apolloscientific.co.uk/product/quinoline-6-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/quinoline-6-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart