APO455832334 Apollo Scientific Ltd

Methyl 5-bromo-1H-indazole-3-carboxylate

  • MDL number MFCD07371572
  • Cas Number : 78155-74-5
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-1H-indazole-3-carboxylate
Inchi Code 1S/C9H7BrN2O2/c1-14-9(13)8-6-4-5(10)2-3-7(6)11-12-8/h2-4H,1H3,(H,11,12)
Inchi Key WVNKCZKRGOOMNU-UHFFFAOYSA-N

Description

Synonym 5-Bromo-3-(methoxycarbonyl)-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-bromo-1h-indazole-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-bromo-1h-indazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart