APO455832335 Apollo Scientific Ltd

Methyl 6-bromo-1H-indazole-3-carboxylate

  • MDL number MFCD07371573
  • Cas Number : 885278-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-3-(methoxycarbonyl)-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-6-bromo-1h-indazole-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-6-bromo-1h-indazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart