APO455832336 Apollo Scientific Ltd

6-Chloro-5-methoxynicotinic acid

  • MDL number MFCD12828186
  • Cas Number : 915107-39-0
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Synonym 6-Chloro-5-methoxypyridine-3-carboxylic acid, 3-Carboxy-6-chloro-5-methoxypyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-5-methoxynicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-5-methoxynicotinic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart