APO455832342 Apollo Scientific Ltd

5-Ethynyl-1H-pyrazolo[4,3-b]pyridine

  • MDL number MFCD22380512
  • Cas Number : 1374652-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Pyrazolo[4,3-b]pyridin-5-yl)acetylene, 5-Ethynyl-4-azaindazole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52987

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52987
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart