APO455832343 Apollo Scientific Ltd

6-Ethynyl-1H-pyrazolo[4,3-c]pyridine

  • MDL number MFCD22380516
  • Cas Number : 1374652-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Pyrazolo[4,3-c]pyridin-6-yl)acetylene, 6-Ethynyl-5-azaindazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52988

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52988
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart