APO455832345 Apollo Scientific Ltd

Methyl 3-methyl-1H-indole-5-carboxylate

  • MDL number MFCD11975645
  • Cas Number : 588688-33-9
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-methyl-1H-indole-5-carboxylate
Inchi Code 1S/C11H11NO2/c1-7-6-12-10-4-3-8(5-9(7)10)11(13)14-2/h3-6,12H,1-2H3
Inchi Key DJVVYXJJXMUAPJ-UHFFFAOYSA-N

Description

Synonym 5-(Methoxycarbonyl)-3-methyl-1H-indole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-methyl-1h-indole-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-methyl-1h-indole-5-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart