APO455832351 Apollo Scientific Ltd

3-[(4,6-Dimethylpyrimidin-2-yl)(methyl)amino]benzoic acid

  • MDL number MFCD00203266
  • Cas Number : 387350-52-9
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 168-170
Iupac Name 3-[(4,6-dimethyl-2-pyrimidinyl)(methyl)amino]benzoic acid
Inchi Code 1S/C14H15N3O2/c1-9-7-10(2)16-14(15-9)17(3)12-6-4-5-11(8-12)13(18)19/h4-8H,1-3H3,(H,18,19)
Inchi Key RBXVYGAOARRNSJ-UHFFFAOYSA-N

Description

Synonym 2-[(3-Carboxyphenyl)(methyl)amino]-4,6-dimethylpyrimidine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 168-170
Product Link https://store.apolloscientific.co.uk/product/3-46-dimethylpyrimidin-2-ylmethylaminobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-46-dimethylpyrimidin-2-ylmethylaminobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart