APO455832354 Apollo Scientific Ltd

4-(Pyrimidin-2-yl)benzoic acid

  • MDL number MFCD01318668
  • Cas Number : 199678-12-1
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 238-240
Iupac Name 4-(2-pyrimidinyl)benzoic acid
Inchi Code 1S/C11H8N2O2/c14-11(15)9-4-2-8(3-5-9)10-12-6-1-7-13-10/h1-7H,(H,14,15)
Inchi Key WNDAEOTYLPWXPN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 238-240
Product Link https://store.apolloscientific.co.uk/product/4-pyrimidin-2-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-pyrimidin-2-ylbenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart