APO455832361 Apollo Scientific Ltd

4-(1H-Imidazol-1-yl)pyridine-2-carboxylic acid

  • MDL number MFCD08056301
  • Cas Number : 914637-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(1H-Imidazol-1-yl)picolinic acid
Purity 97
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-1h-imidazol-1-ylpyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-imidazol-1-ylpyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart