APO455832364 Apollo Scientific Ltd

5-Bromo-2-chloronicotinic acid

  • MDL number MFCD03844847
  • Cas Number : 29241-65-4
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 172-174
Iupac Name 5-bromo-2-chloronicotinic acid
Inchi Code 1S/C6H3BrClNO2/c7-3-1-4(6(10)11)5(8)9-2-3/h1-2H,(H,10,11)
Inchi Key UKNYSJCAGUXDOQ-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-chloropyridine-3-carboxylic acid, 5-Bromo-3-carboxy-2-chloropyridine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 172-174
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloronicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloronicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart