APO455832367 Apollo Scientific Ltd

5-Bromo-6-chloronicotinic acid

  • MDL number MFCD01927098
  • Cas Number : 29241-62-1
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-6-chloropyridine-3-carboxylic acid, 3-Bromo-5-carboxy-2-chloropyridine
Purity 97
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-6-chloronicotinic-acid

Safety Information

signalword Warning
pcodes P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/5-bromo-6-chloronicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart