APO455832371 Apollo Scientific Ltd

5-Methyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD08056309
  • Cas Number : 103879-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Carboxy-5-methyl-1,3-oxazole
Purity 98
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-methyl-13-oxazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-13-oxazole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart