APO455832372 Apollo Scientific Ltd

2-Phenyl-1H-imidazole-4-carboxylic acid hydrate

  • MDL number MFCD04974056
  • Cas Number : 77498-98-7
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 214-216
Iupac Name 2-phenyl-1H-imidazole-4-carboxylic acid hydrate
Inchi Code 1S/C10H8N2O2.H2O/c13-10(14)8-6-11-9(12-8)7-4-2-1-3-5-7;/h1-6H,(H,11,12)(H,13,14);1H2
Inchi Key YIVKWERKOGOQBV-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-2-phenyl-1H-imidazole hydrate, (4-Carboxy-1H-imidazol-2-yl)benzene hydrate
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 214-216
Product Link https://store.apolloscientific.co.uk/product/2-phenyl-1h-imidazole-4-carboxylic-acid-hydrate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-phenyl-1h-imidazole-4-carboxylic-acid-hydrate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart