APO455832374 Apollo Scientific Ltd

2-Amino-4-methylpyrimidine-5-carboxylic acid

  • MDL number MFCD05237187
  • Cas Number : 769-51-7
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4-methyl-5-pyrimidinecarboxylic acid
Inchi Code 1S/C6H7N3O2/c1-3-4(5(10)11)2-8-6(7)9-3/h2H,1H3,(H,10,11)(H2,7,8,9)
Inchi Key DUWNUQBBVGEJPV-UHFFFAOYSA-N

Description

Synonym 5-Carboxy-4-methylpyrimidin-2-amine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-methylpyrimidine-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-methylpyrimidine-5-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart