APO455832379 Apollo Scientific Ltd

6-Chloropyridazine-3-carboxylic acid

  • MDL number MFCD00160464
  • Cas Number : 5096-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-148
Synonym 3-Carboxy-6-chloropyridazine, 3-Carboxy-6-chloro-1,2-diazine
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-chloropyridazine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloropyridazine-3-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart