APO455832380 Apollo Scientific Ltd

6-(Morpholin-4-yl)pyridazine-3-carboxylic acid

  • MDL number MFCD08235211
  • Cas Number : 914637-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-6-(morpholin-4-yl)pyridazine, 4-(6-Carboxypyridazin-3-yl)morpholine
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31785

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31785
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart