APO455832381 Apollo Scientific Ltd

6-(Piperidin-1-yl)pyridazine-3-carboxylic acid

  • MDL number MFCD08235212
  • Cas Number : 914637-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31786

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=31786
Availability details for
Quantity
  • Availability
  • Add to Cart