APO455832382 Apollo Scientific Ltd

2,3-Dihydro-6-methyl-3-oxopyridazine-4-carboxylic acid

  • MDL number MFCD08235213
  • Cas Number : 74557-73-6
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-3-oxo-2,3-dihydro-4-pyridazinecarboxylic acid
Inchi Code 1S/C6H6N2O3/c1-3-2-4(6(10)11)5(9)8-7-3/h2,7H,1H2,(H,8,9)(H,10,11)
Inchi Key GJPNPQDLEXPMPL-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-2,3-dihydro-6-methyl-3-oxopyridazine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-dihydro-6-methyl-3-oxopyridazine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-dihydro-6-methyl-3-oxopyridazine-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart