APO455832387 Apollo Scientific Ltd

1H-Benzimidazole-4-carboxylic acid

  • MDL number MFCD01823426
  • Cas Number : 46006-36-4
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 326-330
Iupac Name 1H-benzimidazole-4-carboxylic acid
Inchi Code 1S/C8H6N2O2/c11-8(12)5-2-1-3-6-7(5)10-4-9-6/h1-4H,(H,9,10)(H,11,12)
Inchi Key VVQNAFBGAWCMLU-UHFFFAOYSA-N

Description

Synonym 1H-Benzimidazole-7-carboxylic acid, 4-Carboxy-1H-benzimidazole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 326-330
Product Link https://store.apolloscientific.co.uk/product/1h-benzimidazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h-benzimidazole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart