APO455832391 Apollo Scientific Ltd

2-Methyl-5-nitro-1H-indole-3-carboxaldehyde

  • MDL number MFCD00216481
  • Cas Number : 3558-17-6
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 305-307(dec.)
Synonym 3-Formyl-2-methyl-5-nitro-1H-indole
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-5-nitro-1h-indole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-5-nitro-1h-indole-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart