APO455832410 Apollo Scientific Ltd

Ethyl 5-amino-3-methylisothiazole-4-carboxylate

  • MDL number MFCD00097916
  • Cas Number : 34859-65-9
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 111-113
Iupac Name ethyl 5-amino-3-methyl-4-isothiazolecarboxylate
Inchi Code 1S/C7H10N2O2S/c1-3-11-7(10)5-4(2)9-12-6(5)8/h3,8H2,1-2H3
Inchi Key ZDGBWOZFPKNORL-UHFFFAOYSA-N

Description

Synonym 5-Amino-4-(ethoxycarbonyl)-3-methylisothiazole, 5-Amino-4-(ethoxycarbonyl)-3-methyl-1,2-thiazole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 111-113
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-amino-3-methylisothiazole-4-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-amino-3-methylisothiazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart