APO455832418 Apollo Scientific Ltd

4-Amino-3-chloropyridine

  • MDL number MFCD03984473
  • Cas Number : 19798-77-7
  • Molecular Weight : 128.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 49-53
Iupac Name 3-chloro-4-pyridinylamine
Inchi Code 1S/C5H5ClN2/c6-4-3-8-2-1-5(4)7/h1-3H,(H2,7,8)
Inchi Key KJBKPRMEMJKXDV-UHFFFAOYSA-N

Description

Synonym 3-Chloropyridin-4-amine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 49-53
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-chloropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-chloropyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart