APO455832424 Apollo Scientific Ltd

2-(Hydroxymethyl)-1-methyl-1H-imidazole

  • MDL number MFCD00964673
  • Cas Number : 17334-08-6
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 107-111
Iupac Name (1-methyl-1H-imidazol-2-yl)methanol
Inchi Code 1S/C5H8N2O/c1-7-3-2-6-5(7)4-8/h2-3,8H,4H2,1H3
Inchi Key CDQDMLWGTVLQEE-UHFFFAOYSA-N

Description

Synonym (1-Methyl-1H-imidazol-2-yl)methanol
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 107-111
Product Link https://store.apolloscientific.co.uk/product/2-hydroxymethyl-1-methyl-1h-imidazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxymethyl-1-methyl-1h-imidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart