APO455832440 Apollo Scientific Ltd

4-Isopropylanisole

  • MDL number MFCD00015033
  • Cas Number : 4132-48-3
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 83
Iupac Name 4-isopropylphenyl methyl ether
density 0.935
Inchi Code 1S/C10H14O/c1-8(2)9-4-6-10(11-3)7-5-9/h4-8H,1-3H3
Inchi Key JULZQKLZSNOEEJ-UHFFFAOYSA-N

Description

Melting Point 83
Synonym 2-(4-Methoxyphenyl)propane, 1-Methoxy-4-(prop-2-yl)benzene, 4-Isopropylphenyl methyl ether, 4-Methoxycumene
Purity 98%
Density 0.935
Flash Point 212-213
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-isopropylanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-isopropylanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart