APO455832447 Apollo Scientific Ltd

3-(4-Iodophenyl)propanoic acid

  • MDL number MFCD00017597
  • Cas Number : 1643-29-4
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 140-142
Iupac Name 3-(4-iodophenyl)propanoic acid
Inchi Code 1S/C9H9IO2/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-2,4-5H,3,6H2,(H,11,12)
Inchi Key SJMRZGIIQIWIFS-UHFFFAOYSA-N

Description

Synonym 4-Iodohydrocinnamic acid
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 140-142
Product Link https://store.apolloscientific.co.uk/product/3-4-iodophenylpropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-iodophenylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart