APO455832450 Apollo Scientific Ltd

7-Bromoheptanoic acid

  • MDL number MFCD00037800
  • Cas Number : 30515-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27-32
Synonym 7-Bromoenanthic acid
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 145/3mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13273

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13273
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart