APO455832457 Apollo Scientific Ltd

5-Chloropentan-1-ol

  • MDL number MFCD00039556
  • Cas Number : 5259-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-5-hydroxypentane, Pentamethylene chlorohydrin
Purity 97
Density 1.06
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 102-103/8mm
Product Link https://store.apolloscientific.co.uk/product/5-chloropentan-1-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloropentan-1-ol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart