APO455832469 Apollo Scientific Ltd

3-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)propanoic acid

  • MDL number MFCD00023096
  • Cas Number : 3339-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-151
Synonym N-(2-Carboxyethyl)phthalimide, 3-(1,3-Dioxoisoindolin-2-yl)propanoic acid
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-13-dihydro-13-dioxo-2h-isoindol-2-ylpropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-13-dihydro-13-dioxo-2h-isoindol-2-ylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335;H302
Availability details for
Quantity
  • Availability
  • Add to Cart