APO455832495 Apollo Scientific Ltd

trans-4-Cyanocinnamic acid

  • MDL number MFCD00016845
  • Cas Number : 16642-94-7
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 245-248
Iupac Name (2E)-3-(4-cyanophenyl)-2-propenoic acid
Inchi Code 1S/C10H7NO2/c11-7-9-3-1-8(2-4-9)5-6-10(12)13/h1-6H,(H,12,13)/b6-5+
Inchi Key USVZQKYCNGNRBV-AATRIKPKSA-N

Description

Synonym (2E)-3-(4-Cyanophenyl)prop-2-enoic acid, (E)-3-(4-Cyanophenyl)acrylic acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 245-248
Product Link https://store.apolloscientific.co.uk/product/trans-4-cyanocinnamic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-4-cyanocinnamic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart