APO455832504 Apollo Scientific Ltd

3'-Acetamidoacetophenone

  • MDL number MFCD00032278
  • Cas Number : 7463-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-130
Synonym 3'-Acetylacetanilide, N-(3-Acetylphenyl)acetamide
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28646

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28646
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart