APO455832507 Apollo Scientific Ltd

2-Chloro-5-iodophenol

  • MDL number MFCD00672969
  • Cas Number : 289039-26-5
  • Molecular Weight : 254.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-iodophenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-iodophenol
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart