APO455832512 Apollo Scientific Ltd

3-Chloropyridine-2-carbonitrile

  • MDL number MFCD03788834
  • Cas Number : 38180-46-0
  • Molecular Weight : 138.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Synonym 3-Chloro-2-cyanopyridine, 3-Chloropicolinonitrile
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloropyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloropyridine-2-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart