APO455832514 Apollo Scientific Ltd

5-Bromo-2-chloronitrobenzene

  • MDL number MFCD05863216
  • Cas Number : 16588-24-2
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 70-72
Iupac Name 4-bromo-1-chloro-2-nitrobenzene
Inchi Code 1S/C6H3BrClNO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H
Inchi Key DJRYWPGOQTUJMQ-UHFFFAOYSA-N

Description

Synonym 4-Bromo-1-chloro-2-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 70-72
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloronitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart