APO455832521 Apollo Scientific Ltd

4-Amino-6-chloropyrimidine-5-carboxaldehyde

  • MDL number MFCD03789626
  • Cas Number : 14160-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 4-Amino-6-chloro-5-formylpyrimidine, 4-Amino-6-chloro-5-formyl-1,3-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-6-chloropyrimidine-5-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-6-chloropyrimidine-5-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart