APO455832524 Apollo Scientific Ltd

3-Chlorophenacyl chloride

  • MDL number MFCD07783975
  • Cas Number : 21886-56-6
  • Molecular Weight : 189.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 40-42
Iupac Name 2-chloro-1-(3-chlorophenyl)ethanone
Inchi Code 1S/C8H6Cl2O/c9-5-8(11)6-2-1-3-7(10)4-6/h1-4H,5H2
Inchi Key AVUVSYIYUADCKE-UHFFFAOYSA-N

Description

Synonym 2,3'-Dichloroacetophenone, 2-Chloro-1-(3-chlorophenyl)ethan-1-one
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 40-42
Product Link https://store.apolloscientific.co.uk/product/3-chlorophenacyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chlorophenacyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart