APO455832526 Apollo Scientific Ltd

4-(Phenylthio)aniline

  • MDL number MFCD00094027
  • Cas Number : 1135-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Phenylsulphanyl)aniline, 4-Aminodiphenyl sulphide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12783

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12783
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart