APO455832539 Apollo Scientific Ltd

5-Bromo-2-chlorobenzaldehyde

  • MDL number MFCD08445659
  • Cas Number : 189628-37-3
  • Molecular Weight : 219.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chlorobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chlorobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart