APO455832554 Apollo Scientific Ltd

4-(Pyrrolidin-1-yl)benzylamine

  • MDL number MFCD04971090
  • Cas Number : 114365-04-7
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-50
Synonym [4-(Pyrrolidin-1-yl)phenyl]methylamine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-pyrrolidin-1-ylbenzylamine

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-pyrrolidin-1-ylbenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart