APO455832557 Apollo Scientific Ltd

2-(1,3-Oxazol-5-yl)aniline

  • MDL number MFCD07772822
  • Cas Number : 774238-36-7
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-56
Synonym 5-(2-Aminophenyl)-1,3-oxazole
Purity 90
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-13-oxazol-5-ylaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-13-oxazol-5-ylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart