APO455832558 Apollo Scientific Ltd

2-Methylimidazo[1,2-a]pyridine-3-carboxaldehyde

  • MDL number MFCD02251327
  • Cas Number : 30384-93-1
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 117-118
Iupac Name 2-methylimidazo[1,2-a]pyridine-3-carbaldehyde
Inchi Code 1S/C9H8N2O/c1-7-8(6-12)11-5-3-2-4-9(11)10-7/h2-6H,1H3
Inchi Key MCSIFRRJDAQKML-UHFFFAOYSA-N

Description

Synonym 3-Formyl-2-methyllimidazo[1,2-a]pyridine
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 117-118
Product Link https://store.apolloscientific.co.uk/product/2-methylimidazo12-apyridine-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylimidazo12-apyridine-3-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart