APO455832560 Apollo Scientific Ltd

2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole

  • MDL number MFCD07772804
  • Cas Number : 92901-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73
Synonym (2,5-Dimethyl-1,3-oxazol-4-yl)methanol
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31302

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31302
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart