APO455832565 Apollo Scientific Ltd

5-Amino-1-(methylsulphonyl)indoline

  • MDL number MFCD01319362
  • Cas Number : 299921-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-117
Synonym 5-Amino-2,3-dihydro-1-(methylsulphonyl)-1H-indole, 2,3-Dihydro-1-(methylsulphonyl)-1H-indol-5-amine
Purity 95
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-methylsulphonylindoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-methylsulphonylindoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart