APO455832569 Apollo Scientific Ltd

4-(Benzyloxy)phenylhydrazine hydrochloride

  • MDL number MFCD00137741
  • Cas Number : 52068-30-1
  • Molecular Weight : 250.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254
Synonym 1-(Benzyloxy)-4-hydrazinobenzene hydrochloride
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxyphenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxyphenylhydrazine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart