APO455832574 Apollo Scientific Ltd

5-Amino-1-methyl-1H-indole

  • MDL number MFCD03839873
  • Cas Number : 102308-97-4
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-104
Synonym 1-Methyl-1H-indol-5-amine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-methyl-1h-indole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-methyl-1h-indole
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart