APO455832589 Apollo Scientific Ltd

1-(2-Phenylethyl)piperazine

  • MDL number MFCD00040740
  • Cas Number : 5321-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-(Piperazin-1-yl)ethyl]benzene
Purity 98
Density 1.006
Flash Point >110
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 276
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15571

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15571
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart